The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014938.5 | NP_055753.3 | 10 - 11 | 1983 | 74 |
XM_006719290.3 | XP_006719353.1 | 10 - 11 | 1967 | 74 | |
XM_006719291.3 | XP_006719354.1 | 10 - 11 | 1967 | 74 | |
XM_006719292.3 | XP_006719355.1 | 10 - 11 | 1967 | 74 | |
XM_006719293.3 | XP_006719356.1 | 10 - 11 | 1967 | 74 | |
XM_006719294.3 | XP_006719357.1 | 4 - 5 | 1026 | 74 | |
GenBank mRNA | AB020674.1 | - | 2 - 3 | 830 | 74 |
AF245480.1 | - | 2 - 3 | 830 | 74 | |
AF245481.1 | - | 2 - 3 | 830 | 74 | |
AF312918.1 | - | 10 - 11 | 1857 | 74 | |
BC069272.1 | - | 5 - 6 | 1120 | 74 | |
BC151841.1 | - | 10 - 11 | 1965 | 74 |