The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003137.4 | NP_003128.3 | 10 - 11 | 1117 | 78 |
NR_034069.1 | 10 - 11 | 1145 | 78 | ||
GenBank mRNA | AJ318054.1 | - | 9 - 10 | 1515 | 78 |
AK292277.1 | - | 10 - 11 | 1062 | 78 | |
AK299591.1 | - | 10 - 11 | 1105 | 78 | |
AK312712.1 | - | 10 - 11 | 1009 | 78 | |
BC038292.1 | - | 10 - 11 | 1082 | 78 | |
BX648663.1 | - | 10 - 11 | 1034 | 78 | |
BX648697.1 | - | 10 - 11 | 1035 | 78 |