The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003137.4 | NP_003128.3 | 5 - 6 | 512 | 158 |
NR_034069.1 | 5 - 6 | 540 | 158 | ||
GenBank mRNA | AJ318054.1 | - | 4 - 5 | 910 | 158 |
AK292277.1 | - | 5 - 6 | 158 | ||
AK299591.1 | - | 5 - 6 | 500 | 158 | |
AK312712.1 | - | 5 - 6 | 404 | 158 | |
BC038292.1 | - | 5 - 6 | 477 | 158 | |
BG772464.1 | - | 5 - 6 | 520 | 158 | |
BX648663.1 | - | 5 - 6 | 429 | 158 | |
BX648697.1 | - | 5 - 6 | 430 | 158 |