The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159291.1 | NP_001152763.1 | 18 - 19 | 2634 | 120 |
NM_001286613.1 | NP_001273542.1 | 19 - 20 | 2928 | 120 | |
NM_022765.3 | NP_073602.3 | 19 - 20 | 2892 | 120 | |
GenBank mRNA | AB048948.1 | - | 19 - 20 | 2892 | 120 |
AK021999.1 | - | 18 - 19 | 2635 | 120 | |
AK024500.1 | - | 3 - 4 | 3657 | 120 | |
AK025392.1 | - | 19 - 20 | 2659 | 120 | |
AK160384.1 | - | 16 - 17 | 3579 | 120 | |
AK225049.1 | - | 19 - 20 | 2649 | 120 | |
AL122098.1 | - | 11 - 12 | 1318 | 120 | |
BC009972.2 | - | 18 - 19 | 2389 | 120 | |
BC042144.1 | - | 19 - 20 | 2788 | 120 | |
BC052983.1 | - | 18 - 19 | 2605 | 120 |