The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159291.1 | NP_001152763.1 | 17 - 18 | 2354 | 85 |
NM_001286613.1 | NP_001273542.1 | 18 - 19 | 2648 | 85 | |
NM_022765.3 | NP_073602.3 | 18 - 19 | 2612 | 85 | |
GenBank mRNA | AB048948.1 | - | 18 - 19 | 2612 | 85 |
AK021999.1 | - | 17 - 18 | 2355 | 85 | |
AK160384.1 | - | 15 - 16 | 3299 | 85 | |
AK225049.1 | - | 18 - 19 | 2369 | 85 | |
AK296284.1 | - | 18 - 19 | 2651 | 85 | |
AL122098.1 | - | 10 - 11 | 1038 | 85 | |
BC009972.2 | - | 17 - 18 | 2109 | 85 | |
BC042144.1 | - | 18 - 19 | 2508 | 85 | |
BC052983.1 | - | 17 - 18 | 2325 | 85 |