The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168551.1 | NP_001162023.1 | 11 - 12 | 1391 | 93 |
NM_018087.4 | NP_060557.3 | 11 - 12 | 1511 | 93 | |
NR_033142.1 | 10 - 11 | 1390 | 93 | ||
XM_011541766.2 | XP_011540068.1 | 11 - 12 | 1353 | 93 | |
GenBank mRNA | AK001269.1 | - | 11 - 12 | 1394 | 93 |
AK295000.1 | - | 11 - 12 | 1175 | 93 | |
AK298909.1 | - | 10 - 11 | 1134 | 93 | |
AK302910.1 | - | 9 - 10 | 969 | 93 | |
AL354612.1 | - | 11 - 12 | 1394 | 93 | |
AL354613.1 | - | 11 - 12 | 1391 | 93 | |
BC003082.2 | - | 11 - 12 | 1461 | 93 | |
BX648031.1 | - | 10 - 11 | 1121 | 93 | |
DQ141696.1 | - | 11 - 12 | 1220 | 93 |