Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UGGT2
Entrez Gene ID
55757
Gene Name
UDP-glucose glycoprotein glucosyltransferase 2
Gene Aliases
HUGT2, UGCGL2, UGT2
Chromosome Location
Chr.13: 95801580 - 96053482 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020121.3 NP_064506.3 10 - 11 1259 103
XM_011521094.1 XP_011519396.1 10 - 11 1214 103
XM_011521095.1 XP_011519397.1 10 - 11 1214 103
XM_011521096.1 XP_011519398.1 10 - 11 1214 103
XM_011521097.1 XP_011519399.1 10 - 11 1214 103
XM_011521098.1 XP_011519400.1 3 - 4 231 103
XM_011521100.2 XP_011519402.1 10 - 11 1214 103
XM_017020662.1 XP_016876151.1 10 - 11 1214 103
XM_017020663.1 XP_016876152.1 11 - 12 1193 103
GenBank mRNA AF227906.2 - 10 - 11 1160 103
AK001735.1 - 10 - 11 1184 103
AL133051.1 - 7 - 8 639 103
BC125233.1 - 10 - 11 1115 103

View Full Product Details