Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UGGT2
Entrez Gene ID
55757
Gene Name
UDP-glucose glycoprotein glucosyltransferase 2
Gene Aliases
HUGT2, UGCGL2, UGT2
Chromosome Location
Chr.13: 95801580 - 96053482 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020121.3 NP_064506.3 13 - 14 1618 113
XM_011521094.1 XP_011519396.1 13 - 14 1573 113
XM_011521095.1 XP_011519397.1 13 - 14 1573 113
XM_011521096.1 XP_011519398.1 13 - 14 1573 113
XM_011521097.1 XP_011519399.1 13 - 14 1573 113
XM_011521098.1 XP_011519400.1 6 - 7 590 113
XM_011521099.2 XP_011519401.1 5 - 6 507 113
XM_011521100.2 XP_011519402.1 13 - 14 1573 113
XM_011521102.2 XP_011519404.1 2 - 3 224 113
XM_011521103.2 XP_011519405.1 3 - 4 335 113
XM_017020662.1 XP_016876151.1 13 - 14 1573 113
XM_017020663.1 XP_016876152.1 14 - 15 1552 113
GenBank mRNA AF227906.2 - 13 - 14 1519 113
AK001735.1 - 13 - 14 1543 113
AL133051.1 - 10 - 11 998 113
BC125233.1 - 13 - 14 1474 113

View Full Product Details