The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020121.3 | NP_064506.3 | 14 - 15 | 1705 | 124 |
XM_011521094.1 | XP_011519396.1 | 14 - 15 | 1660 | 124 | |
XM_011521095.1 | XP_011519397.1 | 14 - 15 | 1660 | 124 | |
XM_011521096.1 | XP_011519398.1 | 14 - 15 | 1660 | 124 | |
XM_011521097.1 | XP_011519399.1 | 14 - 15 | 1660 | 124 | |
XM_011521098.1 | XP_011519400.1 | 7 - 8 | 677 | 124 | |
XM_011521099.2 | XP_011519401.1 | 6 - 7 | 594 | 124 | |
XM_011521100.2 | XP_011519402.1 | 14 - 15 | 1660 | 124 | |
XM_011521102.2 | XP_011519404.1 | 3 - 4 | 311 | 124 | |
XM_011521103.2 | XP_011519405.1 | 4 - 5 | 422 | 124 | |
XM_017020662.1 | XP_016876151.1 | 14 - 15 | 1660 | 124 | |
XM_017020663.1 | XP_016876152.1 | 15 - 16 | 1639 | 124 | |
GenBank mRNA | AF227906.2 | - | 14 - 15 | 1606 | 124 |
AK001735.1 | - | 14 - 15 | 1630 | 124 | |
AL133051.1 | - | 11 - 12 | 1085 | 124 | |
BC125233.1 | - | 14 - 15 | 1561 | 124 |