Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UGGT2
Entrez Gene ID
55757
Gene Name
UDP-glucose glycoprotein glucosyltransferase 2
Gene Aliases
HUGT2, UGCGL2, UGT2
Chromosome Location
Chr.13: 95801580 - 96053482 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020121.3 NP_064506.3 1 - 2 323 140
XM_011521094.1 XP_011519396.1 1 - 2 278 140
XM_011521095.1 XP_011519397.1 1 - 2 278 140
XM_011521096.1 XP_011519398.1 1 - 2 278 140
XM_011521097.1 XP_011519399.1 1 - 2 278 140
XM_011521100.2 XP_011519402.1 1 - 2 278 140
XM_017020662.1 XP_016876151.1 1 - 2 278 140
XM_017020663.1 XP_016876152.1 1 - 2 242 140
GenBank mRNA AF227906.2 - 1 - 2 224 140
AK001735.1 - 1 - 2 248 140
BC017759.1 - 1 - 2 227 140
BC032302.1 - 1 - 2 323 140
BC067807.1 - 1 - 2 234 140
BC111021.1 - 1 - 2 241 140
BC125233.1 - 1 - 2 179 140
BC125234.1 - 1 - 2 179 140

View Full Product Details