Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UGGT2
Entrez Gene ID
55757
Gene Name
UDP-glucose glycoprotein glucosyltransferase 2
Gene Aliases
HUGT2, UGCGL2, UGT2
Chromosome Location
Chr.13: 95801580 - 96053482 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020121.3 NP_064506.3 22 - 23 2810 166
XM_011521094.1 XP_011519396.1 22 - 23 2765 166
XM_011521095.1 XP_011519397.1 22 - 23 2765 166
XM_011521097.1 XP_011519399.1 22 - 23 2765 166
XM_011521098.1 XP_011519400.1 15 - 16 1782 166
XM_011521099.2 XP_011519401.1 14 - 15 1699 166
XM_011521100.2 XP_011519402.1 22 - 23 2765 166
XM_011521102.2 XP_011519404.1 12 - 13 1473 166
XM_011521103.2 XP_011519405.1 12 - 13 1531 166
XM_017020663.1 XP_016876152.1 23 - 24 2748 166
GenBank mRNA AF227906.2 - 22 - 23 2711 166
AK001735.1 - 22 - 23 2735 166
AL133051.1 - 19 - 20 2190 166
BC125233.1 - 22 - 23 2666 166

View Full Product Details