The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020121.3 | NP_064506.3 | 4 - 5 | 657 | 148 |
XM_011521094.1 | XP_011519396.1 | 4 - 5 | 612 | 148 | |
XM_011521095.1 | XP_011519397.1 | 4 - 5 | 612 | 148 | |
XM_011521096.1 | XP_011519398.1 | 4 - 5 | 612 | 148 | |
XM_011521097.1 | XP_011519399.1 | 4 - 5 | 612 | 148 | |
XM_011521100.2 | XP_011519402.1 | 4 - 5 | 612 | 148 | |
XM_017020662.1 | XP_016876151.1 | 4 - 5 | 612 | 148 | |
XM_017020663.1 | XP_016876152.1 | 4 - 5 | 576 | 148 | |
GenBank mRNA | AF227906.2 | - | 4 - 5 | 558 | 148 |
AK001735.1 | - | 4 - 5 | 582 | 148 | |
BC017759.1 | - | 4 - 5 | 561 | 148 | |
BC032302.1 | - | 4 - 5 | 657 | 148 | |
BC067807.1 | - | 4 - 5 | 568 | 148 | |
BC111021.1 | - | 4 - 5 | 575 | 148 | |
BC125233.1 | - | 4 - 5 | 513 | 148 | |
BC125234.1 | - | 4 - 5 | 513 | 148 |