Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL29
Entrez Gene ID
114818
Gene Name
kelch like family member 29
Gene Aliases
KBTBD9
Chromosome Location
Chr.2: 23385419 - 23708613 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052920.1 NP_443152.1 6 - 7 1591 88
XM_006711929.3 XP_006711992.1 5 - 6 2687 88
XM_011532501.2 XP_011530803.1 3 - 4 270 88
XM_017003264.1 XP_016858753.1 5 - 6 1491 88
GenBank mRNA AL834515.1 - 4 - 5 842 88
BC013982.1 - 3 - 4 580 88
BC015667.2 - 2 - 3 603 88

View Full Product Details