The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007553.2 | NP_001007554.1 | 14 - 15 | 2153 | 115 |
NM_001130523.2 | NP_001123995.1 | 14 - 15 | 2207 | 115 | |
NM_001242891.1 | NP_001229820.1 | 15 - 16 | 2300 | 115 | |
NM_001242892.1 | NP_001229821.1 | 13 - 14 | 1766 | 115 | |
NM_001242893.1 | NP_001229822.1 | 12 - 13 | 1673 | 115 | |
NM_007158.5 | NP_009089.4 | 13 - 14 | 2060 | 115 | |
GenBank mRNA | AB020692.1 | - | 14 - 15 | 2067 | 115 |
AF077054.1 | - | 13 - 14 | 1976 | 115 | |
AK022516.1 | - | 12 - 13 | 1575 | 115 | |
AK022528.1 | - | 12 - 13 | 1575 | 115 | |
AK130392.1 | - | 2 - 3 | 261 | 115 | |
AK290146.1 | - | 13 - 14 | 1698 | 115 | |
AL050150.1 | - | 3 - 4 | 353 | 115 | |
AY049788.1 | - | 11 - 12 | 1562 | 115 | |
BC032446.1 | - | 13 - 14 | 1995 | 115 | |
BX648561.1 | - | 13 - 14 | 1897 | 115 | |
BX648780.1 | - | 13 - 14 | 1994 | 115 | |
CR749378.1 | - | 14 - 15 | 2157 | 115 | |
CR749431.1 | - | 14 - 15 | 2139 | 115 |