Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLIP1
Entrez Gene ID
6249
Gene Name
CAP-Gly domain containing linker protein 1
Gene Aliases
CLIP, CLIP-170, CLIP170, CYLN1, RSN
Chromosome Location
Chr.12: 122271432 - 122422956 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001247997.1 NP_001234926.1 21 - 22 3808 124
NM_002956.2 NP_002947.1 20 - 21 3775 124
NM_198240.1 NP_937883.1 19 - 20 3670 124
XM_005253593.2 XP_005253650.1 21 - 22 3871 124
XM_006719552.3 XP_006719615.1 20 - 21 3864 124
XM_017019786.1 XP_016875275.1 20 - 21 3939 124
XM_017019787.1 XP_016875276.1 20 - 21 3754 124
XM_017019788.1 XP_016875277.1 19 - 20 3759 124
XM_017019789.1 XP_016875278.1 19 - 20 3918 124
XM_017019790.1 XP_016875279.1 19 - 20 3660 124
XM_017019791.1 XP_016875280.1 19 - 20 3747 124
XM_017019792.1 XP_016875281.1 18 - 19 3642 124
GenBank mRNA AB209115.1 - 16 - 17 2528 124
AK126990.1 - 10 - 11 1458 124
BC114213.1 - 18 - 19 3585 124
BC117209.1 - 20 - 21 3695 124
BC126305.1 - 20 - 21 3695 124
M97501.1 - 19 - 20 3670 124
X64838.1 - 20 - 21 3752 124

View Full Product Details