Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLIP1
Entrez Gene ID
6249
Gene Name
CAP-Gly domain containing linker protein 1
Gene Aliases
CLIP, CLIP-170, CLIP170, CYLN1, RSN
Chromosome Location
Chr.12: 122271432 - 122422956 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001247997.1 NP_001234926.1 23 - 24 4068 99
NM_002956.2 NP_002947.1 22 - 23 4035 99
NM_198240.1 NP_937883.1 21 - 22 3930 99
XM_005253593.2 XP_005253650.1 23 - 24 4131 99
XM_006719552.3 XP_006719615.1 22 - 23 4124 99
XM_017019786.1 XP_016875275.1 22 - 23 4199 99
XM_017019787.1 XP_016875276.1 22 - 23 4014 99
XM_017019788.1 XP_016875277.1 21 - 22 4019 99
XM_017019789.1 XP_016875278.1 21 - 22 4178 99
XM_017019790.1 XP_016875279.1 21 - 22 3920 99
XM_017019791.1 XP_016875280.1 21 - 22 4007 99
XM_017019792.1 XP_016875281.1 20 - 21 3902 99
GenBank mRNA AB209115.1 - 18 - 19 2788 99
AK123769.1 - 2 - 3 1177 99
AK126990.1 - 12 - 13 1718 99
BC114213.1 - 20 - 21 3845 99
BC117209.1 - 22 - 23 3955 99
BC126305.1 - 22 - 23 3955 99
M97501.1 - 21 - 22 3930 99
X64838.1 - 22 - 23 4012 99

View Full Product Details