The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247997.1 | NP_001234926.1 | 8 - 9 | 1518 | 86 |
XM_005253593.2 | XP_005253650.1 | 8 - 9 | 1581 | 86 | |
XM_017019787.1 | XP_016875276.1 | 8 - 9 | 1581 | 86 | |
GenBank mRNA | AF045653.1 | - | 2 - 3 | 73 | 86 |
BC117209.1 | - | 7 - 8 | 1405 | 86 | |
BC126305.1 | - | 7 - 8 | 1405 | 86 |