Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLIP1
Entrez Gene ID
6249
Gene Name
CAP-Gly domain containing linker protein 1
Gene Aliases
CLIP, CLIP-170, CLIP170, CYLN1, RSN
Chromosome Location
Chr.12: 122271432 - 122422956 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001247997.1 NP_001234926.1 10 - 11 1660 135
NM_002956.2 NP_002947.1 9 - 10 1627 135
XM_005253593.2 XP_005253650.1 10 - 11 1723 135
XM_006719552.3 XP_006719615.1 9 - 10 1716 135
XM_017019786.1 XP_016875275.1 9 - 10 1791 135
XM_017019787.1 XP_016875276.1 10 - 11 1723 135
XM_017019791.1 XP_016875280.1 9 - 10 1716 135
GenBank mRNA AB209115.1 - 6 - 7 725 135
AF045653.1 - 4 - 5 215 135
BC062543.1 - 8 - 9 1583 135
BC117209.1 - 9 - 10 1547 135
BC126305.1 - 9 - 10 1547 135
X64838.1 - 9 - 10 1604 135

View Full Product Details