Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABHD16B
Entrez Gene ID
140701
Gene Name
abhydrolase domain containing 16B
Gene Aliases
C20orf135, dJ591C20.1
Chromosome Location
Chr.20: 63861213 - 63862988 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080622.3 NP_542189.1 1 - 1 112 74
GenBank mRNA AW772131.1 - 1 - 1 112 74

View Full Product Details