The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003404.4 | NP_003395.1 | 3 - 4 | 684 | 79 |
NM_139323.3 | NP_647539.1 | 2 - 3 | 589 | 79 | |
XM_017028039.1 | XP_016883528.1 | 3 - 4 | 488 | 79 | |
GenBank mRNA | AB451260.1 | - | 1 - 2 | 298 | 79 |
AB451386.1 | - | 1 - 2 | 298 | 79 | |
AF064847.1 | - | 3 - 4 | 608 | 79 | |
AK292717.1 | - | 3 - 4 | 577 | 79 | |
BC001359.2 | - | 2 - 3 | 479 | 79 | |
EU794580.1 | - | 3 - 4 | 580 | 79 | |
X57346.1 | - | 3 - 4 | 670 | 79 |