The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136566.2 | NP_001130038.2 | 8 - 9 | 949 | 108 |
XM_006723602.2 | XP_006723665.1 | 8 - 9 | 986 | 108 | |
XM_006723603.2 | XP_006723666.1 | 8 - 9 | 1152 | 108 | |
XM_006723604.2 | XP_006723667.1 | 8 - 9 | 986 | 108 | |
XM_006723605.3 | XP_006723668.1 | 8 - 9 | 986 | 108 | |
XM_011529317.2 | XP_011527619.1 | 8 - 9 | 1148 | 108 | |
XM_011529319.1 | XP_011527621.1 | 6 - 7 | 1018 | 108 | |
XM_011529320.2 | XP_011527622.1 | 6 - 7 | 683 | 108 | |
GenBank mRNA | BC157891.1 | - | 8 - 9 | 949 | 108 |
BC171911.1 | - | 8 - 9 | 949 | 108 | |
HQ603827.1 | - | 7 - 8 | 864 | 108 |