The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159291.1 | NP_001152763.1 | 16 - 17 | 2279 | 88 |
NM_001286613.1 | NP_001273542.1 | 17 - 18 | 2573 | 88 | |
NM_022765.3 | NP_073602.3 | 17 - 18 | 2537 | 88 | |
GenBank mRNA | AB048948.1 | - | 17 - 18 | 2537 | 88 |
AK021999.1 | - | 16 - 17 | 2280 | 88 | |
AK024500.1 | - | 2 - 3 | 2993 | 88 | |
AK025392.1 | - | 17 - 18 | 2304 | 88 | |
AK160384.1 | - | 14 - 15 | 3224 | 88 | |
AK225049.1 | - | 17 - 18 | 2294 | 88 | |
AK296284.1 | - | 17 - 18 | 2576 | 88 | |
AL122098.1 | - | 9 - 10 | 963 | 88 | |
BC009972.2 | - | 16 - 17 | 2034 | 88 | |
BC042144.1 | - | 17 - 18 | 2433 | 88 | |
BC052983.1 | - | 16 - 17 | 2250 | 88 |