The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152729.2 | NP_689942.2 | 5 - 6 | 532 | 176 |
XM_006715378.3 | XP_006715441.1 | 5 - 6 | 535 | 176 | |
GenBank mRNA | AK090918.1 | - | 2 - 3 | 1262 | 176 |
AK290408.1 | - | 5 - 6 | 482 | 176 | |
AK308337.1 | - | 5 - 6 | 515 | 176 | |
AY203935.1 | - | 2 - 3 | 142 | 176 | |
BC015138.1 | - | 4 - 5 | 312 | 175 | |
BC108699.1 | - | 5 - 6 | 489 | 176 | |
BC136832.1 | - | 5 - 6 | 491 | 176 | |
BC136841.1 | - | 5 - 6 | 491 | 176 | |
BM808222.1 | - | - | 521 | 176 |