The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006802.3 | NP_006793.1 | 3 - 4 | 346 | 72 |
GenBank mRNA | AK128438.1 | - | 3 - 4 | 1155 | 72 |
AK222554.1 | - | 3 - 4 | 222 | 72 | |
AK291181.1 | - | 3 - 4 | 227 | 72 | |
BC002395.2 | - | 3 - 4 | 239 | 72 | |
BC011523.1 | - | 3 - 4 | 303 | 72 | |
DA835019.1 | - | 3 - 4 | 346 | 72 | |
U08815.1 | - | 3 - 4 | 209 | 72 |