Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCP2
Entrez Gene ID
6342
Gene Name
sterol carrier protein 2
Gene Aliases
NLTP, NSL-TP, SCP-2, SCP-CHI, SCP-X, SCPX
Chromosome Location
Chr.1: 52927229 - 53051617 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007098.2 NP_001007099.1 1 - 2 241 87
NM_001193599.1 NP_001180528.1 1 - 2 241 87
NM_001193600.1 NP_001180529.1 1 - 2 241 87
NM_002979.4 NP_002970.2 1 - 2 241 87
XM_005271103.4 XP_005271160.1 1 - 2 184 87
XM_011541935.2 XP_011540237.1 1 - 2 184 87
XM_017002045.1 XP_016857534.1 1 - 2 184 87
GenBank mRNA AB208789.1 - 1 - 2 193 87
AK294631.1 - 1 - 2 184 87
AK308105.1 - 1 - 2 184 87
AK312856.1 - 1 - 2 192 87
BC067108.1 - 1 - 2 152 87
BG432929.1 - - 195 87
DC330171.1 - 1 - 2 225 87
M75883.1 - 1 - 2 94 87

View Full Product Details