The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136052.2 | NP_001129524.1 | 18 - 19 | 2415 | 116 |
NM_001199752.2 | NP_001186681.1 | 18 - 19 | 2512 | 116 | |
NM_001199753.1 | NP_001186682.1 | 18 - 19 | 2448 | 116 | |
NM_152359.2 | NP_689572.1 | 17 - 18 | 2379 | 116 | |
NR_108072.1 | 19 - 20 | 2610 | 116 | ||
XM_005258505.2 | XP_005258562.1 | 18 - 19 | 2485 | 116 | |
XM_005258506.4 | XP_005258563.1 | 17 - 18 | 2401 | 116 | |
XM_006723009.3 | XP_006723072.1 | 15 - 16 | 2217 | 116 | |
XM_011526438.2 | XP_011524740.1 | 18 - 19 | 2541 | 116 | |
XM_011526439.2 | XP_011524741.1 | 18 - 19 | 2605 | 116 | |
XM_011526440.2 | XP_011524742.1 | 17 - 18 | 2322 | 116 | |
XM_017026265.1 | XP_016881754.1 | 17 - 18 | 2369 | 116 | |
XM_017026266.1 | XP_016881755.1 | 17 - 18 | 2290 | 116 | |
XM_017026267.1 | XP_016881756.1 | 17 - 18 | 2275 | 116 | |
XM_017026268.1 | XP_016881757.1 | 16 - 17 | 2118 | 116 | |
XM_017026269.1 | XP_016881758.1 | 16 - 17 | 2228 | 116 | |
XM_017026270.1 | XP_016881759.1 | 16 - 17 | 1996 | 116 | |
XM_017026271.1 | XP_016881760.1 | 16 - 17 | 2085 | 116 | |
XM_017026272.1 | XP_016881761.1 | 16 - 17 | 2177 | 116 | |
XM_017026273.1 | XP_016881762.1 | 16 - 17 | 2162 | 116 | |
GenBank mRNA | AF357970.1 | - | 17 - 18 | 2337 | 116 |
AK074389.1 | - | 10 - 11 | 1358 | 116 | |
AK096516.1 | - | 19 - 20 | 2606 | 116 | |
AK289713.1 | - | 18 - 19 | 2504 | 116 | |
AK290092.1 | - | 18 - 19 | 2420 | 116 | |
BC029104.1 | - | 18 - 19 | 2379 | 116 |