Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPT1C
Entrez Gene ID
126129
Gene Name
carnitine palmitoyltransferase 1C
Gene Aliases
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
Chromosome Location
Chr.19: 49690672 - 49713731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136052.2 NP_001129524.1 14 - 15 1847 61
NM_001199752.2 NP_001186681.1 14 - 15 1944 61
NM_001199753.1 NP_001186682.1 14 - 15 1880 61
NM_152359.2 NP_689572.1 13 - 14 1811 61
NR_108072.1 15 - 16 2042 61
XM_005258505.2 XP_005258562.1 14 - 15 1917 61
XM_005258506.4 XP_005258563.1 13 - 14 1833 61
XM_006723009.3 XP_006723072.1 11 - 12 1649 61
XM_011526438.2 XP_011524740.1 14 - 15 1973 61
XM_011526439.2 XP_011524741.1 14 - 15 2037 61
XM_011526440.2 XP_011524742.1 14 - 15 1889 61
XM_017026265.1 XP_016881754.1 14 - 15 1936 61
XM_017026266.1 XP_016881755.1 14 - 15 1857 61
XM_017026267.1 XP_016881756.1 13 - 14 1707 61
XM_017026268.1 XP_016881757.1 12 - 13 1550 61
XM_017026269.1 XP_016881758.1 12 - 13 1660 61
XM_017026270.1 XP_016881759.1 12 - 13 1428 61
XM_017026271.1 XP_016881760.1 12 - 13 1517 61
XM_017026272.1 XP_016881761.1 13 - 14 1744 61
XM_017026273.1 XP_016881762.1 13 - 14 1729 61
GenBank mRNA AB208884.1 - 8 - 9 924 61
AF357970.1 - 13 - 14 1769 61
AK074389.1 - 6 - 7 790 61
AK096516.1 - 15 - 16 2038 61
AK289713.1 - 14 - 15 1936 61
AK290092.1 - 14 - 15 1852 61
AK299866.1 - 11 - 12 1638 61
AK309087.1 - 13 - 14 1770 61
AL831876.1 - 4 - 5 442 61
BC029104.1 - 14 - 15 1811 61

View Full Product Details