The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136052.2 | NP_001129524.1 | 8 - 9 | 1088 | 68 |
XM_017026266.1 | XP_016881755.1 | 8 - 9 | 1098 | 68 | |
XM_017026271.1 | XP_016881760.1 | 6 - 7 | 758 | 68 | |
GenBank mRNA | BC029104.1 | - | 8 - 9 | 1052 | 68 |