Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED25
Entrez Gene ID
81857
Gene Name
mediator complex subunit 25
Gene Aliases
ACID1, ARC92, BVSYS, CMT2B2, P78, PTOV2, TCBAP0758
Chromosome Location
Chr.19: 49818279 - 49840384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011527353.1 XP_011525655.1 17 - 18 2202 67
GenBank mRNA AF283769.2 - 12 - 13 1670 67
AY533507.1 - - 2238 67

View Full Product Details