The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282985.1 | NP_001269914.1 | 2 - 3 | 414 | 88 |
NM_025195.3 | NP_079471.1 | 2 - 3 | 1240 | 88 | |
GenBank mRNA | AF205437.1 | - | 2 - 3 | 931 | 88 |
AF250310.1 | - | 2 - 3 | 939 | 88 | |
AJ000480.1 | - | 1 - 2 | 214 | 88 | |
AK297539.1 | - | 2 - 3 | 355 | 88 | |
AL832388.1 | - | 2 - 3 | 872 | 88 | |
BC063292.1 | - | 2 - 3 | 1240 | 88 | |
DA686993.1 | - | 2 - 3 | 414 | 88 |