The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005513.2 | NP_005504.2 | 2 - 3 | 563 | 119 |
XM_011512744.2 | XP_011511046.1 | 2 - 3 | 686 | 119 | |
XM_011512745.2 | XP_011511047.1 | 2 - 3 | 634 | 119 | |
GenBank mRNA | AK223401.1 | - | 2 - 3 | 529 | 119 |
S67859.1 | - | 2 - 3 | 512 | 119 | |
X63468.1 | - | 1 - 2 | 503 | 119 |