Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCOLCE2
Entrez Gene ID
26577
Gene Name
procollagen C-endopeptidase enhancer 2
Gene Aliases
PCPE2
Chromosome Location
Chr.3: 142817860 - 142889203 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013363.3 NP_037495.1 8 - 9 1423 78
GenBank mRNA AF098269.1 - 8 - 9 1149 78
AK075478.1 - 8 - 9 78
AK223633.1 - 8 - 9 1345 78
AK315121.1 - 8 - 9 1191 78
AY035400.1 - 8 - 9 1312 78
AY358557.1 - 8 - 9 1422 78
BC006265.2 - 8 - 9 1176 78
CR536570.1 - 8 - 9 1116 78

View Full Product Details