Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POU2F2
Entrez Gene ID
5452
Gene Name
POU class 2 homeobox 2
Gene Aliases
OCT2, OTF2, Oct-2
Chromosome Location
Chr.19: 42086110 - 42197931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207025.2 NP_001193954.1 10 - 11 1011 66
NM_001207026.1 NP_001193955.1 10 - 11 1011 66
NM_001247994.1 NP_001234923.1 10 - 11 963 66
NM_002698.4 NP_002689.1 10 - 11 963 66
XM_005259010.3 XP_005259067.1 10 - 11 980 66
XM_011527040.2 XP_011525342.1 10 - 11 1094 66
XM_011527041.2 XP_011525343.1 10 - 11 1094 66
XM_011527042.2 XP_011525344.1 10 - 11 1046 66
XM_011527043.2 XP_011525345.2 11 - 12 1391 66
XM_017026884.1 XP_016882373.1 11 - 12 1392 66
XM_017026885.1 XP_016882374.1 11 - 12 1395 66
XM_017026886.1 XP_016882375.1 12 - 13 2894 66
XM_017026887.1 XP_016882376.1 12 - 13 1364 66
XM_017026888.1 XP_016882377.1 12 - 13 1844 66
XM_017026889.1 XP_016882378.1 11 - 12 1386 66
XM_017026890.1 XP_016882379.1 11 - 12 1467 66
XM_017026891.1 XP_016882380.1 11 - 12 1344 66
XM_017026892.1 XP_016882381.1 11 - 12 1326 66
XM_017026893.1 XP_016882382.1 10 - 11 1046 66
XM_017026894.1 XP_016882383.1 10 - 11 1028 66
XM_017026895.1 XP_016882384.1 10 - 11 1026 66
XM_017026896.1 XP_016882385.1 11 - 12 1392 66
GenBank mRNA BC006101.1 - 10 - 11 955 66
BT007438.1 - 10 - 11 901 66
M22596.1 - 2 - 3 187 66
M36542.1 - 10 - 11 955 66
M36653.1 - 10 - 11 1015 66
M36718.1 - 10 - 11 946 66
X13809.1 - 10 - 11 946 66
X13810.1 - 10 - 11 955 66
X53468.1 - 10 - 11 1015 66

View Full Product Details