Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POU2F2
Entrez Gene ID
5452
Gene Name
POU class 2 homeobox 2
Gene Aliases
OCT2, OTF2, Oct-2
Chromosome Location
Chr.19: 42086110 - 42197931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207025.2 NP_001193954.1 13 - 14 1463 75
NM_001247994.1 NP_001234923.1 13 - 14 1279 75
NM_002698.4 NP_002689.1 13 - 14 1415 75
XM_011527041.2 XP_011525343.1 13 - 14 1546 75
XM_011527042.2 XP_011525344.1 13 - 14 1498 75
XM_011527043.2 XP_011525345.2 14 - 15 1707 75
XM_017026896.1 XP_016882385.1 14 - 15 1844 75
GenBank mRNA BC006101.1 - 13 - 14 1271 75
M36542.1 - 13 - 14 1407 75
M36718.1 - 13 - 14 1398 75
M36772.1 - 1 - 2 54 75
X13809.1 - 13 - 14 1398 75
X13810.1 - 13 - 14 1407 75
X53469.1 - 1 - 2 54 75

View Full Product Details