The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022114.3 | NP_071397.3 | 12 - 13 | 3017 | 76 |
NM_199454.2 | NP_955533.2 | 12 - 13 | 3017 | 76 | |
XM_005244772.4 | XP_005244829.1 | 12 - 13 | 3169 | 76 | |
XM_005244773.4 | XP_005244830.1 | 12 - 13 | 3173 | 76 | |
XM_005244774.4 | XP_005244831.1 | 12 - 13 | 3172 | 76 | |
XM_006710814.3 | XP_006710877.1 | 12 - 13 | 3167 | 76 | |
XM_011541945.2 | XP_011540247.1 | 11 - 12 | 2790 | 76 | |
XM_017002050.1 | XP_016857539.1 | 12 - 13 | 3017 | 76 | |
GenBank mRNA | AB051462.2 | - | 12 - 13 | 2967 | 76 |
AB078876.1 | - | 12 - 13 | 2979 | 76 | |
AF294278.1 | - | 12 - 13 | 2998 | 76 | |
AK309078.1 | - | 11 - 12 | 2713 | 76 | |
BC161614.1 | - | 12 - 13 | 2967 | 76 |