The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024419.4 | NP_077733.3 | 8 - 9 | 1597 | 106 |
NR_110601.1 | 7 - 8 | 1446 | 106 | ||
NR_110602.1 | 7 - 8 | 1481 | 106 | ||
NR_111989.1 | 7 - 8 | 1349 | 106 | ||
XM_011525487.2 | XP_011523789.1 | 8 - 9 | 1972 | 106 | |
XM_011525489.2 | XP_011523791.1 | 8 - 9 | 1899 | 106 | |
XM_017025357.1 | XP_016880846.1 | 8 - 9 | 1972 | 106 | |
XM_017025358.1 | XP_016880847.1 | 8 - 9 | 1972 | 106 | |
XM_017025361.1 | XP_016880850.1 | 8 - 9 | 1591 | 106 | |
XM_017025362.1 | XP_016880851.1 | 8 - 9 | 1591 | 106 | |
XM_017025363.1 | XP_016880852.1 | 8 - 9 | 1543 | 106 | |
XM_017025364.1 | XP_016880853.1 | 7 - 8 | 1493 | 106 | |
XM_017025365.1 | XP_016880854.1 | 7 - 8 | 1445 | 106 | |
GenBank mRNA | AK024529.1 | - | 7 - 8 | 1318 | 106 |
AK225030.1 | - | 7 - 8 | 1318 | 106 | |
AK296882.1 | - | 8 - 9 | 1571 | 106 | |
AK297156.1 | - | 7 - 8 | 1437 | 106 | |
AK309890.1 | - | 4 - 5 | 1098 | 106 | |
AK316147.1 | - | 8 - 9 | 1575 | 106 | |
AL359590.1 | - | 3 - 4 | 694 | 106 | |
BC025951.1 | - | 8 - 9 | 1530 | 106 | |
BC035662.1 | - | 7 - 8 | 1406 | 106 | |
BC108732.1 | - | 8 - 9 | 1561 | 106 | |
CR749720.1 | - | 3 - 4 | 694 | 106 |