Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL36G
Entrez Gene ID
56300
Gene Name
interleukin 36, gamma
Gene Aliases
IL-1F9, IL-1H1, IL-1RP2, IL1E, IL1F9, IL1H1, IL1RP2
Chromosome Location
Chr.2: 112978019 - 112985672 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019618.3 NP_062564.1 2 - 3 133 90
GenBank mRNA AF200492.1 - 2 - 3 128 90
AF206696.1 - 2 - 3 120 90
BC096721.1 - 1 - 2 78 90
BC098130.1 - 1 - 2 78 90
BC098155.1 - 1 - 2 78 90
BC098337.1 - 1 - 2 78 90

View Full Product Details