The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018071.3 | NP_001018081.3 | 18 - 19 | 2690 | 65 |
NM_001318191.1 | NP_001305120.1 | 16 - 17 | 2331 | 65 | |
GenBank mRNA | AK123038.1 | - | 16 - 17 | 2331 | 65 |
AK131386.1 | - | 16 - 17 | 2176 | 65 | |
AK303923.1 | - | 18 - 19 | 2388 | 65 | |
CR749241.1 | - | 18 - 19 | 2662 | 65 |