The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018071.3 | NP_001018081.3 | 19 - 20 | 2801 | 116 |
NM_001318191.1 | NP_001305120.1 | 17 - 18 | 2442 | 116 | |
GenBank mRNA | AK123038.1 | - | 17 - 18 | 2442 | 116 |
AK131386.1 | - | 17 - 18 | 2287 | 116 | |
AK303923.1 | - | 19 - 20 | 2499 | 116 | |
CR749241.1 | - | 19 - 20 | 116 |