The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243372.1 | NP_001230301.1 | 4 - 5 | 975 | 82 |
NM_001243374.1 | NP_001230303.1 | 3 - 4 | 643 | 82 | |
NM_001829.3 | NP_001820.2 | 4 - 5 | 975 | 82 | |
NM_173872.3 | NP_776297.2 | 4 - 5 | 975 | 82 | |
XM_005262726.2 | XP_005262783.1 | 3 - 4 | 515 | 82 | |
XM_011531586.2 | XP_011529888.1 | 3 - 4 | 1025 | 82 | |
GenBank mRNA | AB019542.1 | - | 4 - 5 | 503 | 82 |
AF029346.1 | - | 3 - 4 | 416 | 82 | |
AF172729.1 | - | 3 - 4 | 416 | 82 | |
AK127483.1 | - | 3 - 4 | 643 | 82 | |
AK304362.1 | - | 4 - 5 | 909 | 82 | |
BC054877.1 | - | 4 - 5 | 833 | 82 | |
BC136510.1 | - | 5 - 6 | 750 | 82 | |
BP284718.1 | - | 3 - 4 | 498 | 82 | |
BX647119.1 | - | 4 - 5 | 925 | 82 | |
X78520.1 | - | 4 - 5 | 904 | 82 |