Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SALL3
Entrez Gene ID
27164
Gene Name
spalt like transcription factor 3
Gene Aliases
ZNF796
Chromosome Location
Chr.18: 78980275 - 78998969 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171999.3 NP_741996.2 1 - 2 83 87
GenBank mRNA BC148296.2 - 1 - 2 83 87

View Full Product Details