Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RERGL
Entrez Gene ID
79785
Gene Name
RERG like
Gene Aliases
-
Chromosome Location
Chr.12: 18080869 - 18090193 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286201.1 NP_001273130.1 1 - 2 104 122
NR_104413.1 1 - 2 104 122
GenBank mRNA AA220227.1 - 1 - 2 52 122
BX422875.2 - 1 - 2 52 122

View Full Product Details