Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRSS12
Entrez Gene ID
8492
Gene Name
protease, serine 12
Gene Aliases
BSSP-3, BSSP3, MRT1
Chromosome Location
Chr.4: 118280038 - 118353042 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003619.3 NP_003610.2 12 - 13 2365 83
GenBank mRNA AF077298.1 - 3 - 4 476 83
AJ001531.1 - 12 - 13 2358 83

View Full Product Details