The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005969.3 | NP_005960.1 | 15 - 16 | 1302 | 97 |
GenBank mRNA | BC022090.2 | - | 15 - 16 | 1212 | 97 |
U77456.1 | - | 15 - 16 | 1310 | 97 |