The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016653.2 | NP_057737.2 | 6 - 7 | 648 | 86 |
NM_133646.2 | NP_598407.1 | 6 - 7 | 648 | 86 | |
XM_005246640.2 | XP_005246697.1 | 6 - 7 | 694 | 86 | |
XM_017004323.1 | XP_016859812.1 | 6 - 7 | 694 | 86 | |
XM_017004324.1 | XP_016859813.1 | 6 - 7 | 718 | 86 | |
GenBank mRNA | AB030034.1 | - | 6 - 7 | 623 | 86 |
AB041883.1 | - | 6 - 7 | 718 | 86 | |
AB049733.1 | - | 5 - 6 | 448 | 86 | |
AB049734.1 | - | 5 - 6 | 86 | ||
AB208974.1 | - | 6 - 7 | 86 | ||
AF238255.1 | - | 5 - 6 | 448 | 86 | |
AF251441.1 | - | 6 - 7 | 86 | ||
AF325454.1 | - | 5 - 6 | 86 | ||
AF465843.1 | - | 6 - 7 | 617 | 86 | |
AF480461.1 | - | 6 - 7 | 86 | ||
AF480462.1 | - | 6 - 7 | 643 | 86 | |
AK056310.1 | - | 6 - 7 | 649 | 86 | |
AK291825.1 | - | 6 - 7 | 540 | 86 | |
AK298634.1 | - | 5 - 6 | 413 | 86 | |
BC001401.2 | - | 6 - 7 | 578 | 86 |