The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278352.1 | NP_001265281.1 | 7 - 8 | 885 | 63 |
NM_002197.2 | NP_002188.1 | 6 - 7 | 813 | 63 | |
XM_005251476.1 | XP_005251533.1 | 7 - 8 | 823 | 63 | |
XM_011517888.1 | XP_011516190.1 | 6 - 7 | 735 | 63 | |
GenBank mRNA | AB209480.1 | - | 6 - 7 | 768 | 63 |
AF261088.1 | - | 7 - 8 | 864 | 63 | |
AK057904.1 | - | 6 - 7 | 781 | 63 | |
AK308752.1 | - | 5 - 6 | 779 | 63 | |
BC018103.1 | - | 5 - 6 | 695 | 63 | |
EU794627.1 | - | 5 - 6 | 695 | 63 | |
M58510.1 | - | 4 - 5 | 441 | 63 | |
Z11559.1 | - | 6 - 7 | 765 | 63 |