The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278352.1 | NP_001265281.1 | 9 - 10 | 1195 | 122 |
NM_002197.2 | NP_002188.1 | 8 - 9 | 1123 | 122 | |
XM_005251476.1 | XP_005251533.1 | 9 - 10 | 1133 | 122 | |
XM_011517888.1 | XP_011516190.1 | 8 - 9 | 1045 | 122 | |
GenBank mRNA | AB209480.1 | - | 8 - 9 | 1078 | 122 |
AF261088.1 | - | 9 - 10 | 1174 | 122 | |
AK057904.1 | - | 8 - 9 | 1091 | 122 | |
AK308752.1 | - | 7 - 8 | 1089 | 122 | |
BC018103.1 | - | 7 - 8 | 1005 | 122 | |
EU794627.1 | - | 7 - 8 | 1005 | 122 | |
M58510.1 | - | 6 - 7 | 751 | 122 | |
Z11559.1 | - | 8 - 9 | 1075 | 122 |