The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199054.1 | NP_001185983.1 | 2 - 3 | 238 | 60 |
NM_001199055.1 | NP_001185984.1 | 2 - 3 | 206 | 60 | |
NM_001199056.1 | NP_001185985.1 | 2 - 3 | 206 | 60 | |
NM_013399.2 | NP_037531.2 | 2 - 3 | 206 | 60 | |
GenBank mRNA | AF131218.1 | - | 1 - 2 | 285 | 60 |
AF195661.1 | - | 2 - 3 | 201 | 60 | |
AK292036.1 | - | 2 - 3 | 164 | 60 | |
AK294257.1 | - | 2 - 3 | 176 | 60 | |
AK302297.1 | - | 2 - 3 | 164 | 60 | |
AL136698.1 | - | 2 - 3 | 164 | 60 | |
AL833853.1 | - | 2 - 3 | 238 | 60 | |
BC002882.2 | - | 2 - 3 | 152 | 60 | |
DQ167023.1 | - | 2 - 3 | 175 | 60 |