The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152244.1 | NP_689450.1 | 1 - 2 | 280 | 103 |
GenBank mRNA | AK023932.1 | - | 1 - 2 | 280 | 103 |
AK296569.1 | - | 1 - 2 | 262 | 103 |