Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX11
Entrez Gene ID
29916
Gene Name
sorting nexin 11
Gene Aliases
-
Chromosome Location
Chr.17: 48107533 - 48123064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013323.2 NP_037455.2 1 - 2 250 99
XM_005257261.3 XP_005257318.1 1 - 2 258 99
GenBank mRNA AF121861.1 - 1 - 2 270 99
AK091852.1 - 2 - 3 235 99

View Full Product Details